missing translation for 'onlineSavingsMsg'
Learn More

Atrazine 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30327375
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327375 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327375 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011094AC
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atrazine
CAS 1912-24-9
Analyte or Component Names Atrazine
Shipping Condition Ice pack (-20°C)
Molecular Formula C8 H14 Cl N5
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-ethyl-N4-(1-methylethyl)-, 1,3,5-Triazine-2,4-diamine, 6-chloro-N-ethyl-N'-(1-methylethyl)- (9CI), s-Triazine, 2-chloro-4-(ethylamino)-6-(isopropylamino)- (8CI), 6-Chloro-N2-ethyl-N4-(1-methylethyl)-1,3,5-triazine-2,4-diamine, 1-Chloro-3-ethylamino-5-isopropylamino-2,4,6-triazine, 2-Chloro-4-(ethylamino)-6-(2-propylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)-s-triazine, 2-Chloro-4-(ethylamino)-6-(isopropylamino)triazine, 2-Chloro-4-ethylamineisopropylamine-s-triazine, 2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-1,3,5-triazine, 2-Ethylamino-4-isopropylamino-6-chloro-s-triazine, 6-Chloro-4-(ethylamino)-2-(isopropylamino)-s-triazine, 6-Chloro-N-Ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diamine, A 361, AAtrex, AAtrex 4L, AAtrex Nine-O, ATZ, Aatrax, Aatrex 4LC, Aatrex L, Akticon, Aktikon, Aktikon PK, Aktinit A, Aktinit PK, Argezin, Atradex, Atragranz, Atranex, Atranex 80WP, Atraphyt, Atrasan, Atrataf, Atrazerba FL, Atrazin, Atrazine, Azoprim, Azoprim 50, CET, Chromozin, Cyazin, Farmozine, G 30027, Gesamprim, Gesaprim, Gesaprim 50, Gesaprim 500, Gesaprim L, Herbatoxol, Hungazin, Hungazin PK, NSC 163046, Nortox 500SC, Nu-Trazine, Oleogesaprim, Oleogesaprim 200, Primatol A, Primoleo, Radazin, Radazin T, Triazine A 1294, Wonuk, Zealin L, Zeapos, Zeazin, Zeazine, Zeopos
InChI Formula InChI=1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)
SMILES CCNc1nc(Cl)nc(NC(C)C)n1
IUPAC Name 6-chloro-4-N-ethyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine
Solvent or Matrix Acetone
Molecular Weight (g/mol) 215.68
Quantity 1 mL
Formula Weight 215.0938 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.