missing translation for 'onlineSavingsMsg'
Learn More

Atropic Acid, TRC

Product Code. 30133345
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133345 1 g 1g
17911945 250 mg 250mg
17940465 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133345 Supplier Toronto Research Chemicals Supplier No. TRCA7938501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-38-6
Molecular Formula C9 H8 O2
Molecular Weight (g/mol) 148.16
Synonym 2-Phenylpropenoic acid, Atropic acid, Ipratropium Bromide Imp. D (EP), Benzeneacetic acid, α-methylene-, Atropic acid (6CI,7CI,8CI), α-Methylenebenzeneacetic acid, 2-Phenylacrylic acid, 2-Phenylprop-2-enoic acid, 2-Propenoic acid, 2-phenyl-, NSC 20987, α-Phenylacrylic acid
IUPAC Name 2-phenylprop-2-enoic acid
SMILES OC(=O)C(=C)c1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8O2/c1-7(9(10)11)8-5-3-2-4-6-8/h2-6H,1H2,(H,10,11)
Formula Weight 148.0524
Chemical Name or Material Atropic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.