missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATRX / RAD54 Mouse, Clone: 39f, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 546-MSM1-P1
This item is not returnable.
View return policy
Specifications
| ATRX / RAD54 | |
| Nuclear | |
| 39f | |
| Unconjugated | |
| ATRX | |
| Recombinant human ATRX protein | |
| Raji, HeLa or A431 Cells. Pancreas, Adrenal, Ovary, Kidney, Prostate or Stomach. | |
| 100 μg | |
| Primary | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| Mouse | |
| 280kDa | |
| Protein A/G Purified | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction