Learn More
3M™ Attest™ Vaporized Hydrogen Peroxide Tri-Metric Chemical Indicator Strips
Product Code. 17741679
Change view
Click to view available options
Quantity:
25 Strips/Pouch, 4 Pouches/Cs.
Unit Size:
Pack of 250
Product Code. Quantity Unit Size
17741679 25 Strips/Pouch, 4 Pouches/Cs. Pack of 250
1 options
Product Code. 17741679 Supplier 3M™ Supplier No. 7100227991
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Verify that the stated values for exposure time, temperature, and amount of vaporized hydrogen peroxide have been achieved within a package or containment device and/or at a specific location within the load or empty chamber.
Upon exposure to vaporized hydrogen peroxide, the color of the coated indicator strip progressively changes from blue toward pink. The progression of the color change along the strip is visible through a window with marked 'REJECT' and 'ACCEPT' zones.
  • The first FDA cleared CI for VH2O2 sterilization, categorized by the FDA as a multivariable CI for chemical vapor sterilization that independently monitors the three critical variables of time, temperature and VH2O2 concentration
  • Accurately detects failed VH2O2 sterilization cycles
  • Won't change color before sterilization under ambient light
  • Consistent blue-toward-pink color change for all 3M™ chemical indicators for vaporized hydrogen peroxide
  • Proprietary protective film prevents ink smear on surgical instruments
Specifications
Certifications/Compliance FDA VH2O2 Sterilization
For Use With (Equipment) Wrapped Trays, Rigid Containers, Sterilization Pouches, Packs
Length (English) 2 in.
Length (Metric) 5.1 cm
Model 1348
Product Type Indicator Strip
Quantity 25 Strips/Pouch, 4 Pouches/Cs.
Sterility Sterile