Learn More
Isofield™ Atto 230 Cleanroom Irradiated Tie-On Facemask with Anti Fog Visor - Grade A/B Compatible
BIOPHARMA
Product Code. 30178870
Change view
Click to view available options
Size:
Wide
Unit Size:
Pack of 200
Product Code. Size Unit Size
30178870 Wide Pack of 200
1 options
Product Code. 30178870 Supplier Isofield™ Supplier No. 60521
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Gamma irradiated visor facemask with long length ties threaded through the visor to ensure a close fit to user. The Atto is made from low-linting cleanroom grade material. The Atto has excellent bacterial and particle filtration efficiency, with an anti-scratch and anti-fog visor.
  • Irradiated tie-on cleanroom facemask with anti-fog visor
  • 3-layer non-woven configuration
  • Threaded top ties to ensure secure fit to face
  • Anti-fog-coated visor
  • Highly efficient bacterial and particle filtration
  • Particle Shedding (Helmke Drum) Category I
  • Individually packed pouches printed with IPA resistant ink to maintain traceability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Polyethylene, Polypropylene
Size Wide
Sterility Sterile
Style Pleated