missing translation for 'onlineSavingsMsg'
Learn More

Atuveciclib S-Enantiomer, MedChemExpress

Product Code. 16478137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478137 5 mg 5mg
16435428 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478137 Supplier Medchem Express Supplier No. HY12871C
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Atuveciclib S-Enantiomer (BAY-1143572 S-Enantiomer) is a potent and selective CDK9 inhibitor, which inhibits CDK9/CycT1 with an IC50 of 16 nM.

Atuveciclib S-Enantiomer (BAY-1143572 S-Enantiomer) is a potent and selective CDK9 inhibitor, which inhibits CDK9/CycT1 with an IC50 of 16 nM.

Specifications

Chemical Name or Material Atuveciclib S-Enantiomer
CAS 250279-81-1
Color White
Purity Grade Notes Research
Molecular Formula C18H18FN5O2S
Quantity 5 mg
Synonym BAY-1143572 S-Enantiomer
Solubility Information DMSO : ≥ 113 mg/mL (291.67 mM)
SMILES N=[S@@](CC1=CC(NC2=NC(C3=CC=C(F)C=C3OC)=NC=N2)=CC=C1)(C)=O
Molecular Weight (g/mol) 387.43
Formula Weight 387.43
Percent Purity 99.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.