missing translation for 'onlineSavingsMsg'
Learn More

3M™ Aura™ Particulate Respirators 9300+ Series

Product Code. 11717574 Shop All 3M Products
Change view
Click to view available options
Filter Type:
FFP1
FFP2
FFP3
With Exhalation Valve:
No
Yes
Assigned Protection Factor (APF):
4
10
20
Model:
9310
9312
9320
9322+
9322+ P2 valved
9330
9330+ P3
9332+ P3 valved
PF9312
PF9320
PF9322
PF9332
Unit Size:
Pack of 10
Pack of 20
Pack of 330
Pack of 440
Pack of 5
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type With Exhalation Valve Assigned Protection Factor (APF) Model Unit Size
11717574 FFP1 No 4 9310 Pack of 20
10583051 FFP2 No 10 9320 Pack of 20
10116544 FFP2 Yes 10 9322+ Pack of 10
12672545 FFP1 Yes 4 9312 Pack of 10
12602555 FFP3 No 20 9330 Pack of 20
11777574 FFP1 Yes 4 PF9312 Pack of 5
11787574 FFP2 No 10 PF9320 Pack of 5
11797574 FFP2 Yes 10 PF9322 Pack of 5
11707584 FFP3 Yes 20 PF9332 Pack of 5
16523771 FFP3 Yes 20 9332+ P3 valved Pack of 330
16684912 FFP2 Yes 10 9322+ P2 valved Pack of 330
16674902 FFP3 No 20 9330+ P3 Pack of 440
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 11717574 Supplier 3M™ Supplier No. 7000088725

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Particulate respirators have an ergonomic, flat-fold, 3-panel design featuring a low breathing resistance filter media. Available in FFP1, FFP2, and FFP3 protection levels with or without valves, Innovative chin tab for easy adjustments and an embossed, sculpted upper panel helps reduce eyewear fogging.

  • Provide comfortable protection against dusts and mists in a wide variety of industrial applications and other hazardous environments.
  • Offers FFP1, FFP2 and FFP3 protection.
  • 3-panel design allows for greater facial movement.
  • Sculpted, low-profile style of the embossed upper panel creates an effective seal, reduces eyewear fogging and improves eyewear compatibility.
  • Low resistance filter technology promotes easier breathing and the innovative chin tab allows simple adjustments.
  • Choose without valve style or with valve styles that feature 3M™ Cool Flow™ to reduce heat build-up and help provide comfortable protection in hot and humid conditions.
  • Individually packed in hygienic wrapping to prevent contamination before use.
  • Braided, colour-coded headbands help you identify the protection level offered: yellow for FFP1 (APF 4), blue for FFP2 (APF 10) and red for FFP3 (APF 20).
  • Foldable design makes them easy to store when not in use.
  • Respirators work well with 3M eyewear and hearing protection products.
TRUSTED_SUSTAINABILITY

Specifications

Assigned Protection Factor (APF) 4
Certifications/Compliance CE Approved to EN 149:2001A1:2009
Certifications/Compliance (Europe) EN 149:2001 + A1:2009
Certifications (EN) EN 149:2001 + A1:2009
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP1
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve No
Filtration Layer Material Polypropylene
For Use With (Application) Agriculture, Cleaning Maintenance, Demolition
Latex Free Yes
Material Headstrap Polyisoprene
Model 9310
Nose Piece Material Aluminum, Polyurethene
Nosepiece Type Sculpted
Packaging Type Individually Packaged
Quantity 20/Pk.
Resistant To Dusts and mists found in a wide variety of industrial applications and other work situations
Shape Cup
Sterility Non-sterile
Style 3-panel
Strap Attachment Type Colour-coded Headband, Yellow
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.