missing translation for 'onlineSavingsMsg'
Learn More

3M™ Aura™ Particulate Respirator 06900+Gen3 Series

Product Code. 16280252
Change view
Click to view available options
Filter Efficiency:
FFP2
FFP3
With Exhalation Valve:
Yes
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Efficiency With Exhalation Valve unitSize
16280252 FFP3 Yes Pack of 10
16270252 FFP2 Yes Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16280252 Supplier 3M™ Supplier No. 7100134423

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Aura™ Particulate Respirator 06900+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.

  • Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
  • New 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort
  • 3M Advanced Electret (filter) Media helps you breathe easier
  • Innovative tabs on the upper and lower panels simplify positioning and adjustment
  • Braided headbands are color-coded for quick and easy identification
  • Easy to store away with flat-fold design and individual wrapping
  • Breathing protection against hazardous work environments
  • Made in the UK
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Approved; EN 149:2001+A1:2009 FFP2 NR D
Product Type Flat Fold
With Exhalation Valve Yes
Filter Efficiency FFP3
Model 6933+Gen3
Quantity 10/Pk.
Length (Metric) 242 mm
Width (Metric) 87 mm
Height (Metric) 212 mm
Weight (Metric) 304 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.