Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Brand:  3M™ 7000088836
Code : S7
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12942505
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Provide reliable and comfortable protection from a range of airborne disposable hazards
  • Minimize contamination risks to food or pharmaceutical production processes
  • Made without staples or small detachable parts
  • Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear
Specifications
Specifications
FFP3