missing translation for 'onlineSavingsMsg'
Learn More

Aurothioglucose, MedChemExpress

Product Code. 16490718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490718 5 mg 5mg
16468288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490718 Supplier Medchem Express Supplier No. HYA0068
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aurothioglucose (Gold thioglucose) is a well known active-site inhibitor of TrxR1, inhibited TrxR1 activity in HeLa cell cytosol but had no effect on the viability of the cells.

Aurothioglucose (Gold thioglucose) is a well known active-site inhibitor of TrxR1, inhibited TrxR1 activity in HeLa cell cytosol but had no effect on the viability of the cells.

Especificaciones

Chemical Name or Material Aurothioglucose
Physical Form Powder
CAS 12192-57-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H11AuO5S
For Use With (Application) Cancer-programmed cell death
Synonym Gold thioglucose
Solubility Information H2O : 125 mg/mL (318.73 mM; Need ultrasonic) ∣DMSO : 6 mg/mL (15.30 mM; Need ultrasonic)
SMILES O[C@@H]1[C@](O[Au+][SH-]2)([H])C2O[C@H](CO)[C@H]1O
Molecular Weight (g/mol) 392.18
Formula Weight 392.18
Grade Research
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.