missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Autoclave Bags
Designed for contaminated waste disposal in autoclaves or incinerators.
Brand: Microspec™ H5
Additional Details : Weight : 3.40000kg
Description
- Clear bags imprinted with blue biohazard labeling designed for safe disposal
- Constructed of thick polypropylene
- Allows for repeated autoclaving
- Strong side seams for securely holding items
- Front write-on panel
Specifications
Autoclave Bags | |
Autoclavable | |
Single-use | |
Non-sterile | |
Polypropylene | |
EcoPak | |
Medium | |
632 mm | |
413 mm | |
62.733 lb. |
Room Temperature | |
Clear with Blue Print | |
Polypropylene | |
50 μm | |
200 | |
200 | |
2 mil | |
16.26 in. | |
413 mm | |
28.455 kg |