missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Autoclave Bags
Designed for contaminated waste disposal in autoclaves or incinerators.
32.30 € - 297.00 €
Specifications
Product Type | Autoclave Bags |
---|---|
Content And Storage | Room Temperature |
Autoclavable | Autoclavable |
Color | Clear with Blue Print |
Disposable | Single-use |
Product Code | Brand | Quantity | Length (Metric) | Outer Packaging | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Length (Metric) | Outer Packaging | Price | Quantity & Availability | |||
15702135
|
Microspec™
G1 |
1500 | 302 mm | EcoPak |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15712135
|
Microspec™
G5 |
200 | 302 mm | Bag |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15722135
|
Microspec™
H1 |
1500 | 632 mm | EcoPak |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15732135
|
Microspec™
H5 |
200 | 632 mm | EcoPak |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15742135
|
Microspec™
I1 |
1200 | 633 mm | EcoPak |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15752135
|
Microspec™
I5 |
200 | 633 mm | Bag |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Clear bags imprinted with blue biohazard labeling designed for safe disposal
- Constructed of thick polypropylene
- Allows for repeated autoclaving
- Strong side seams for securely holding items
- Front write-on panel
Specifications
Autoclave Bags | |
Autoclavable | |
Single-use | |
Non-sterile | |
Polypropylene |
Room Temperature | |
Clear with Blue Print | |
Polypropylene | |
50 μm | |
2 mil |