Learn More
Novus Biologicals™ Autophagy Antibody Pack
For Research applications
Brand: Novus Biologicals™ NB910-94877
Includes:
- 1.NB100-2220: LC3B/MAP1LC3B Antibody
- 2.NB110-53818: ATG5 Antibody
- 3.NB110-56893: ATG9A Antibody
- 4.NB110-82384: ATG16L1 Antibody
- 5.NB110-87318: Beclin 1/ATG6 Antibody
Description
Specifications
| Autophagy | |
| 25ul vials of each of following: NB100-2220, NB110-87318, NB110-53818, NB110-56893 and NB110-82384. | |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated | |
| ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 | |
| See individual datasheets | |
| Autophagy | |
| Autophagy Antibodies (Beclin, LC3, ATG5, ATG9, ATG16L1) | |
| Human, Mouse, Rat, Pig, Alligator |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| See individual datasheets | |
| BECN1 | |
| 5 Vials | |
| 8678 | |
| 1.NB100-2220: LC3B/MAP1LC3B Antibody 2.NB110-53818: ATG5 Antibody 3.NB110-56893: ATG9A Antibody 4.NB110-82384: ATG16L1 Antibody 5.NB110-87318: Beclin 1/ATG6 Antibody |
|
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |