missing translation for 'onlineSavingsMsg'
Learn More

Auxinole, MedChemExpress

Product Code. 16470767
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470767 50 mg 50mg
16467177 5 mg 5mg
16465336 100 mg 100mg
16424458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470767 Supplier Medchem Express Supplier No. HY111444
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Auxinole is a potent auxin antagonist of TIR1/AFB receptors, binding TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression.

Auxinole is a potent auxin antagonist of TIR1/AFB receptors, binding TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression.

Specifications

Chemical Name or Material Auxinole
Physical Form Solid
CAS 86445-22-9
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H19NO3
Solubility Information DMSO : ≥ 125 mg/mL (388.96 mM) ∣Ethanol : 2.5 mg/mL (7.78 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)C(CC(C1=CC=C(C)C=C1C)=O)C2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 321.37
Formula Weight 321.37
Percent Purity 99.19%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.