missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Avelumab

Product Code. 16491109
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491109 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16491109 Supplier MedChemExpress Supplier No. HY108730

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avelumab is a fully human IgG1 anti-PD-L1 monoclonal antibody with potential antibody-dependent cell-mediated cytotoxicity.

Specifications

Antigen PD-1/PD-L1
Quantity 1 mg
Target Species Human
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Product Type Inhibitory Antibodies
Shipping Condition Room temperature
CAS 1537032-82-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.