missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avian Influenza NS1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 € - 507.00 €
Specifications
| Antigen | Avian Influenza NS1 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18688235
|
Novus Biologicals
NBP2-41067-0.025mg |
0.025 mg |
178.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18117877
|
Novus Biologicals
NBP2-41067 |
0.1 mg |
507.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Avian Influenza NS1 Polyclonal antibody specifically detects Avian Influenza NS1 in Virus samples. It is validated for ELISA.Specifications
| Avian Influenza NS1 | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the carboxy terminus of the Avian Influenza Nonstructural Protein 1 protein. Efforts were made to use relatively conserved regions of the viral sequence as the antigen. The immunogen is located within the last 50 amino acids of Avian Influenza Nonstructural Protein 1. Amino Acid Squence: LPPNQKRKMARTIE | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title