Learn More
AXIN1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1C4E8, Proteintech
Mouse Monoclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | AXIN1 |
|---|---|
| Clone | 1C4E8 |
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18045275
|
Proteintech
CL488-68093-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030496
|
Proteintech
CL488-68093-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The AXIN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AXIN1. This antibody recognizes human, mouse, rat antigen. The AXIN1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| AXIN1 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 8312 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1C4E8 | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| AXIN, axin 1, AXIN1, Axis inhibition protein 1, hAxin | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.