missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well Rigid PCR Microplate, Non-sterile
GREENER_CHOICE

Product Code. 15531433 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15531433 Clear Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15531433 Supplier Axygen™ Supplier No. PCR96HSACC
  • ONLINE EXCLUSIVE PRICE
    226.00 €
    223.43 € / Pack of 50
    Save 2.57 €
    1% Off
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

96-well half skirt automation compatible PCR microplate

Axygen™ automation-compatible PCR microplate features a rigid frame with a small split that sustains heat transferring during and post thermal cycling without distorting its shape. The polypropylene plate also features raised rims around the wells to provide tight sealing to minimize heat evaporation. Compatible with most major thermal cyclers, sequencers, and automation platforms, the plate provides optimal performance for downstream applications, including high throughput screening.
  • Remains flat during and post thermal cycling
  • Raised well rims help reduce heat evaporation
  • Clear wells, visible grid for easy sample loading and tracking
  • Ultra-thin and consistent wall thickness for easy and consistent heat transfer
  • Suitable for standard and real-time PCR
  • Single notch corner
  • SBS footprint
  • Manufactured from polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Non-sterile
  • Compatible with major automation platforms, thermal cyclers, and sequencers
TRUSTED_SUSTAINABILITY

Specifications

Array 8 x 12
Color Clear
For Use With (Equipment) Automation compatible
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
No. per Case 50/Cs.
No. per Pack 10/Pk.
RNase Free Yes
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.