missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile
GREENER_CHOICE

Product Code. p-4102745 Shop All Axygen Scientific Products
Click to view available options
No. of Wells:
24
32
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 12789408

Brand: Axygen™ PCR24C

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to fit a wide variety of thermal cyclers

  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Lid Without Lid
Array 4 x 6
Color Clear
Product Type PCR Microplate, without Lid
Material Polypropylene
No. of Wells 24
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.