missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates

Product Code. 10068469 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Orange
Red
Violet
White
Yellow
Volume (Metric) Well:
40 μL
50 μL
55 μL
Includes:
Double notch
Single notch
Unit Size:
Pack of 100
Pack of 20
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Volume (Metric) Well Includes unitSize
10068469 White 55 μL Single notch Pack of 50
12719418 Blue - Double notch Pack of 50
12749418 Clear 50 μL Single notch Pack of 50
12759418 Green - Double notch Pack of 50
12709438 Orange - Double notch Pack of 50
12739438 Clear 50 μL Double notch Pack of 100
12567817 Clear 50 μL Double notch Pack of 20
12719438 Red 40 μL Double notch Pack of 50
12759438 Violet 40 μL Double notch Pack of 50
12769438 Yellow - Double notch Pack of 50
15328301 Clear 50 μL Single notch Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10068469 Supplier Axygen™ Supplier No. PCR384LC480WNF
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Single and double notch 384-well PCR plates

Sealing Options: Axymats, heat sealing, adhesive films and foils.

  • Available in 50μL and 55μL
  • Single and double notch corner

Skirted (50μL)

  • Application/Compatibility: standard thermal cycling
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted (55μL)
  • Application/Compatibility: standard thermal cycling, fully ABI compatible for qPCR and sequencing
  • Robotic-friendly full skirt design
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.
Skirted, RigidPlate (50μL)
  • Robotic-friendly rigid plate with full skirt
  • Application/Compatibility: standard thermal cycling, ABI sequencing, automation applications
  • Packaging Description: 10 plates/Pk., 2 packs/sleeve, 5 sleeves/Cs.
Skirted, elevated wells
  • Application/Compatibility: standard thermal cycling, ABI sequencing, qPCR
  • Raised wells allow 55μL volume
  • Packaging Description: 10 plates/Pk., 5 packs/Cs.

Colors available by special order.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance RNase, DNase-free
Format Standard
Lid Without Lid
Array 16 x 24
Color White
For Use With (Equipment) For Roche light cycler 480
Material Polypropylene
Volume (Metric) Well 55 μL
No. of Wells 384
Volume (Metric) Working 30 μL
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
Includes Single notch
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.