missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates

Product Code. 12771206 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
Product Code. Color unitSize
12771206 White Pack of 50
1 options
This item is not returnable. View return policy

Product Code. 12771206

Brand: Axygen™ PCR384LC480W

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

RNase-/DNase-free

Axygen™ 384 well PCR microplate, white, is designed to fit Roche Light Cycler 480. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Includes Ultra Clear sealing film and printed alphanumeric grid for ease of well identification.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Includes Sealing Film
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Certifications/Compliance RNase, DNase-free
    Format Standard
    Lid Without Lid
    Description Sealing film
    Array 16 x 24
    Color White
    For Use With (Equipment) For Roche light cycler 480
    Skirt Style Full
    Material Polypropylene
    Volume (Metric) Well 55 μL
    No. of Wells 384
    Volume (Metric) Working 30 μL
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR
    Includes Single notch
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Well Color White
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.