missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 96-Well Half-Skirt PCR Microplates
Shop All Axygen Scientific Products
Click to view available options
Color:
Red
White
Yellow
Unit Size:
Pack of 50
Description
PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.
- 96-well PCR half-skirt amplification plate with a single notch
- Working volume is 300μL
- Available in a variety of colors
- Sold 10 plates/unit; 5 units/Cs.
Specifications
Specifications
| Certifications/Compliance | RNase, DNase-free |
| Format | Standard |
| Array | 8 x 12 |
| Color | Red |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Round |
| For Use With (Application) | PCR thermal transfer, RT-PCR applications |
| Quantity | 50/Cs. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction