missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well Low Profile PCR Microplates

Product Code. 12794706 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
White
Skirt Style:
Half
Non-skirt
Unit Size:
Pack of 100
Pack of 25
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style unitSize
12794706 Clear Non-skirt Pack of 100
12774706 Clear Half Pack of 100
11370195 Clear Half Pack of 25
12759468 White Half Pack of 100
12789468 White Non-skirt Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12794706 Supplier Axygen™ Supplier No. PCR96LPFLTC
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The low profile design results in faster and more efficient PCR reactions.

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications. The

Axygen™ 96 well low profile no skirt, flat top PCR microplate is designed to fit low profile 96 well blocks. Low profile design results in faster and more efficient PCR reactions. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Suitable for 0.1 mL thermal cycler blocks
  • 96-Well Flat top low profile PCR plate
  • Application/Compatibility: Standard Biometra thermal cyclers, Eppendorf MasterCycler qPCR, MJ Opticon/Chromo4 qPCR
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode No Barcode
    Certifications/Compliance RNase-/DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color Clear
    Product Type Flat top
    For Use With (Equipment) 0.1 mL thermal cycler blocks; Standard Biometra thermal cyclers, Eppendorf MasterCycler qPCR, MJ Opticon/Chromo4 qPCR
    Skirt Style Non-skirt
    Material Polypropylene
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.