missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well 100μL PCR Microplates

Product Code. 10834221 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10834221 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10834221 Supplier Axygen™ Supplier No. PCR96LC480WNF
Estimated Shipment: 07-12-2026
Aggiungi al carrello
Aggiungi al carrello
Bulk Quote Request

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well 100μL PCR Microplates are specifically designed to be compatible with Roche Light Cycler 480.

Axygen™ 96 well PCR microplate is designed to fit Roche Light Cycler 480. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Printed alphanumeric grid for ease of well identification.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • TRUSTED_SUSTAINABILITY

    Specifica

    Barcode No Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White
    Product Type PCR Microplate
    For Use With (Equipment) Roche Light Cycler 480
    Material Polypropylene
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Includes Printed alphanumeric grid
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Vedi altri risultati Mostra meno risultati
    Titolo del prodotto
    Selezionare un problema

    Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.