missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well 100μL PCR Microplates

Product Code. 10743262 Shop All Axygen Scientific Products
Change view
Click to view available options
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Barcode Unit Size
10743262 Includes Barcode Pack of 50
10352200 No Barcode Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10743262 Supplier Axygen™ Supplier No. PCR96LC480WBC
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well 100μL PCR Microplates are specifically designed to be compatible with Roche Light Cycler 480.

Axygen™ 96 well PCR microplate is designed to fit Roche Light Cycler 480. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Includes Ultra Clear sealing film and printed alphanumeric grid for ease of well identification.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • Includes Sealing Films
  • TRUSTED_SUSTAINABILITY

    Specifications

    Barcode Includes Barcode
    Certifications/Compliance RNase-, DNase-free
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Color White
    Product Type PCR Microplate
    For Use With (Equipment) Roche Light Cycler 480
    Material Polypropylene
    Volume (Metric) Well 100 μL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Includes Ultra Clear sealing film, bar codes, printed alphanumeric grid
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.