missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Storage Microplates
BIOPHARMAGREENER_CHOICE

Product Code. 12597927 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Blue
Clear
Green
Sterility:
Non-sterile
Sterile
No. of Wells:
24
48
96
384
Volume (Metric) Well:
1.1 mL
1.8 mL
10,000 μL
1100 μL
125 μL
2000 μL
240 μL
5000 μL
550 μL
Unit Size:
Pack of 25
Pack of 30
Pack of 50
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility No. of Wells Volume (Metric) Well unitSize
12597927 Clear Non-sterile 96 1.8 mL Pack of 50
12547907 Clear Non-sterile 384 125 μL Pack of 50
12557907 Clear Sterile 384 125 μL Pack of 50
12587907 Clear Sterile 384 240 μL Pack of 50
11340115 Clear Sterile 48 5000 μL Pack of 25
11386074 Clear Non-sterile 384 240 μL Pack of 50
11330115 Clear Non-sterile 48 5000 μL Pack of 25
12547917 Green Non-sterile 96 550 μL Pack of 50
12587917 Blue Non-sterile 96 550 μL Pack of 50
12587927 Clear Sterile 96 1100 μL Pack of 50
12557837 Clear Sterile 24 10,000 μL Pack of 25
12577927 Clear Non-sterile 96 1.1 mL Pack of 50
12537837 Clear Non-sterile 24 10,000 μL Pack of 25
11350135 Clear Sterile 96 2000 μL Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 12597927 Supplier Axygen™ Supplier No. PDW20C
Powering Proteomics
Use promo code 28003
View all promotions
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

For sample collection, in-vitro growth chambers and long term storage.

Used for sample collection, in vitro growth chambers and long-term storage.

Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.


  • Excellent chemical resistance
  • ANSI/SLAS 1-2004: Microplates - Footprint Dimensions
  • Centrifugation to 4,000 RCF
  • Diamond shaped wells allow complete sample recovery
  • Black and white plates are available by special order
  • Ultra-flat surface ensures proper sealing with heat sealing film
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance ANSI, SLAS 1-2004
    Format Standard
    Lid Without Lid
    Array 8 x 12
    Autoclavable Autoclavable
    Color Clear
    Material Polypropylene
    Volume (Metric) Well 1.8 mL
    No. of Wells 96
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Max. RCF 4000 x g
    No. per Case 50/Cs.
    No. per Pack 5/Pk.
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.