Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Product Code. 12627215 Shop All Axygen Scientific Products
Change view
Click to view available options
Pre-slit:
No
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pre-slit Sterility Unit Size
12627215 No Non-sterile Pack of 50
12551328 No Sterile Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12627215 Supplier Axygen™ Supplier No. AM500ULRD
  • ONLINE EXCLUSIVE PRICE
    340.00 €
    336.27 € / Pack of 50
    Save 3.73 €
    1% Off
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Case 50/Cs.
No. per Pack 10/Pk.
Pre-slit No
Product Type Sealing Mat
Well Shape Round
Plate Compatibility 96-well Deep Well Plate, 0.5 mL