missing translation for 'onlineSavingsMsg'
Learn More

Azacyclonol, MedChemExpress

Product Code. 16480978
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480978 5 g 5g
16462219 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480978 Supplier Medchem Express Supplier No. HYB0530
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Azacyclonol (γ-pipradol), a metabolite of Terfenadine, is a central depressant agent. Azacyclonol is a ganglion-blocking agent. Azacyclonol can be used to diminish psychoses-induced hallucinations.

Azacyclonol (γ-pipradol), a metabolite of Terfenadine, is a central depressant agent. Azacyclonol is a ganglion-blocking agent. Azacyclonol can be used to diminish psychoses-induced hallucinations.

Specifications

Chemical Name or Material Azacyclonol
Physical Form Solid
CAS 115-46-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H21NO
For Use With (Application) Neuroscience-Neuromodulation
Synonym γ-pipradol
Solubility Information DMSO : 100 mg/mL (374.01 mM; Need ultrasonic)
SMILES OC(C1=CC=CC=C1)(C2CCNCC2)C3=CC=CC=C3
Molecular Weight (g/mol) 267.37
Formula Weight 267.37
Percent Purity 99.99%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.