missing translation for 'onlineSavingsMsg'
Learn More

Azaperone, MedChemExpress

Product Code. 16498766
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498766 100 mg 100mg
16482276 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498766 Supplier Medchem Express Supplier No. HYB1470
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azaperone (R-1929) acts as a dopamine antagonist but also has some antihistaminic and anticholinergic properties. Azaperone is a pyridinylpiperazine and butyrophenone neuroleptic drug with antiemetic effects, which is used mainly as a tranquilizer in veterinary medicine.

Azaperone (R-1929) acts as a dopamine antagonist but also has some antihistaminic and anticholinergic properties. Azaperone is a pyridinylpiperazine and butyrophenone neuroleptic drug with antiemetic effects, which is used mainly as a tranquilizer in veterinary medicine.

Specifications

Chemical Name or Material Azaperone
CAS 1649-18-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H22FN3O
Quantity 100 mg
Synonym R-1929
Solubility Information DMSO : 50 mg/mL (152.72 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.05 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(F)C=C1)CCCN2CCN(C3=NC=CC=C3)CC2
Molecular Weight (g/mol) 327.4
Formula Weight 327.4
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.