missing translation for 'onlineSavingsMsg'
Learn More

AZD2906, MedChemExpress

Product Code. 16483236
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483236 25 mg 25mg
16465586 100 mg 100mg
16457527 5 mg 5mg
16424788 10 mg 10mg
16411057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16483236 Supplier Medchem Express Supplier No. HY113854
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

Specifications

Chemical Name or Material AZD2906
CAS 1034148-15-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H25FN4O3
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (271.44 mM; Need ultrasonic)
SMILES FC1=CC=C(N2N=CC3=CC(O[C@@H]([C@@H](NC(C4CC4)=O)C)C5=CC=C(N=C5)OC)=CC=C23)C=C1
Molecular Weight (g/mol) 460.5
Formula Weight 460.5
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.