missing translation for 'onlineSavingsMsg'
Learn More

AZD7545, MedChemExpress

Product Code. 16476978
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476978 10 mg 10mg
16447056 100 mg 100mg
16443037 50 mg 50mg
16429527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476978 Supplier Medchem Express Supplier No. HY16082
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD7545 is a potent, competitive, selective PDHK2 (pyruvate dehydrogenase kinase 2) inhibitor with IC50s of 36.8 nM, 6.4 nM for PDHK1 and PDHK2, respectively.

AZD7545 is a potent, competitive, selective PDHK2 (pyruvate dehydrogenase kinase 2) inhibitor with IC50s of 36.8 nM, 6.4 nM for PDHK1 and PDHK2, respectively.

Specifications

Chemical Name or Material AZD7545
CAS 252017-04-2
Color White
Purity Grade Notes Research
Molecular Formula C19H18ClF3N2O5S
Quantity 10 mg
Solubility Information DMSO : ≥ 46 mg/mL (96.06 mM)
SMILES O=C(N(C)C)C1=CC=C(S(=O)(C2=CC=C(NC([C@@](C)(O)C(F)(F)F)=O)C(Cl)=C2)=O)C=C1
Molecular Weight (g/mol) 478.87
Formula Weight 478.87
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.