missing translation for 'onlineSavingsMsg'
Learn More

AZD8186, MedChemExpress

Product Code. 16467817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467817 5 mg 5mg
16435098 10 mg 10mg
16425826 100 mg 100mg
16401337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467817 Supplier Medchem Express Supplier No. HY12330
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD8186 is a PI3K inhibitor, which potently inhibits PI3Kβ (IC50=4 nM) and PI3Kδ (IC5050=12 nM) with selectivity over PI3Kα (IC50=35 nM) and PI3Kγ (IC50=675 nM).

AZD8186 is a PI3K inhibitor, which potently inhibits PI3Kβ (IC50=4 nM) and PI3Kδ (IC5050=12 nM) with selectivity over PI3Kα (IC50=35 nM) and PI3Kγ (IC50=675 nM).

Specifications

Chemical Name or Material AZD8186
CAS 1627494-13-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H25F2N3O4
Quantity 5 mg
Solubility Information DMSO : ≥ 35 mg/mL (76.51 mM)
SMILES O=C(C1=CC([C@H](NC2=CC(F)=CC(F)=C2)C)=C3C(C(C=C(N4CCOCC4)O3)=O)=C1)N(C)C
Molecular Weight (g/mol) 457.47
Formula Weight 457.47
Percent Purity 99.67%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.