missing translation for 'onlineSavingsMsg'
Learn More

AZD8330, MedChemExpress

Product Code. 16484958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484958 10 mg 10mg
16461207 50 mg 50mg
16457697 5 mg 5mg
16445736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484958 Supplier Medchem Express Supplier No. HY12058
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD8330 (ARRY-424704) is a potent, uncompetitive MEK1/MEK2 inhibitor, with an IC50 of 7 nM.

AZD8330 (ARRY-424704) is a potent, uncompetitive MEK1/MEK2 inhibitor, with an IC50 of 7 nM.

Specifications

Chemical Name or Material AZD8330
CAS 869357-68-6
Color Pink
Purity Grade Notes Research
Molecular Formula C16H17FIN3O4
Quantity 10 mg
Synonym ARRY-424704 ARRY-704
Solubility Information DMSO : ≥ 100 mg/mL (216.81 mM)
SMILES IC1=CC(F)=C(C=C1)NC(N2C)=C(C=C(C2=O)C)C(NOCCO)=O
Molecular Weight (g/mol) 461.23
Formula Weight 461.23
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.