missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azilsartan-d5, MedChemExpress
Description
Azilsartan D5 (TAK-536 D5) is the deuterium labeled Azilsartan(TAK-536), which is a specific and potent angiotensin II type 1 receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Azilsartan-d5 |
| CAS | 1346599-45-8 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C25H15D5N4O5 |
| Color | White |
| Synonym | TAK-536-d5 |
| SMILES | OC(C1=CC=CC2=C1N(C(OC(C([2H])([2H])[2H])([2H])[2H])=N2)CC(C=C3)=CC=C3C4=CC=CC=C4C5=NC(ON5)=O)=O |
| Molecular Weight (g/mol) | 461.48 |
| Quantity | 1 mg |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?