missing translation for 'onlineSavingsMsg'
Learn More

Azimsulfuron, TRC

Product Code. 30133791
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133791 50 mg 50mg
17904505 25 mg 25mg
17902855 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133791 Supplier Toronto Research Chemicals Supplier No. TRCA29014550MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120162-55-2
Molecular Formula C13 H16 N10 O5 S
Molecular Weight (g/mol) 424.4
Synonym N-[[(4,6-Dimethoxy-2-pyrimidinyl)amino]carbonyl]-1-methyl-4-(2-methyl-2H-tetrazol-5-yl)-1H-pyrazole-5-sulfonamide, Azimsulfuron, DPX 47, DPX-A 8947, Gulliver, IN-A 8947
IUPAC Name 1-(4,6-dimethoxypyrimidin-2-yl)-3-[2-methyl-4-(2-methyltetrazol-5-yl)pyrazol-3-yl]sulfonylurea
SMILES COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2c(cnn2C)c3nnn(C)n3)n1

Specifications

Analyte or Component Names Azimsulfuron
Shipping Condition Room Temperature
InChI Formula InChI=1S/C13H16N10O5S/c1-22-11(7(6-14-22)10-18-21-23(2)19-10)29(25,26)20-13(24)17-12-15-8(27-3)5-9(16-12)28-4/h5-6H,1-4H3,(H2,15,16,17,20,24)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 50 mg
Formula Weight 424.1026
Percent Purity >95%
Chemical Name or Material Azimsulfuron
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.