missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Lids for sterilizing trough

Product Code. 12073974
Change view
Click to view available options
For Use With (Equipment):
10 L Sterilizing Trough
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12073974 10 L Sterilizing Trough Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12073974 Supplier Azlon Supplier No. TWR205
  • ONLINE EXCLUSIVE PRICE
    36.54 €
    30.15 € / Pack of 1
    Save 6.39 €
    17% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with sterilizing trough

  • Lids are to be purchased separately from trough
  • Lids are not autoclavable

Specifications

Autoclavable Not Autoclavable
Color Transparent
For Use With (Equipment) 10 L Sterilizing Trough
Material Polystyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.