missing translation for 'onlineSavingsMsg'
Learn More

Azodicarbonamide, 97%

Product Code. 10589620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10589620 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10589620 Supplier Thermo Scientific Acros Supplier No. 105165000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 123-77-3 | C2H4N4O2 | 116.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-77-3
Molecular Weight (g/mol) 116.08
MDL Number MFCD00007958
InChI Key XOZUGNYVDXMRKW-AATRIKPKSA-N
Synonym azodicarbonamide, azodicarboxamide, diazenedicarboxamide, azobisformamide, azodiformamide, azodicarbamide, azodicarboamide, azobiscarbonamide, azobiscarboxamide, celogen az
PubChem CID 5462814
IUPAC Name (E)-carbamoyliminourea
SMILES C(=O)(N)N=NC(=O)N

Specifications

Melting Point 195.0°C to 202.0°C
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C2H4N4O2
Linear Formula H2NCON=NCONH2
Quantity 500 g
Beilstein 03,IV,246
Merck Index 15,912
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in common solvents, soluble in dimethyl sulfoxide, slightly soluble in demethylformamide
Formula Weight 116.08
Percent Purity 97%
Chemical Name or Material Azodicarbonamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Azodicarbonamide
Signal Word
  • Danger
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.