accessibility menu, dialog, popup

UserName

NH2ONNNH2O(E/Z)

CAS RN 123-77-3

IUPAC Name: (carbamoylimino)urea
Synonyms: diazenedicarboxamide
Molecular Weight (g/mol): 116.08
Molecular Formula: C2H4N4O2
InChi Key: XOZUGNYVDXMRKW-UHFFFAOYSA-N
SMILES: NC(=O)N=NC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results