missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aztreonam

Product Code. 15585146 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585146 1 g 1g
15595146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15585146 Supplier Thermo Scientific Chemicals Supplier No. 455410010
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Type Aztreonam
Melting Point 227.0°C
Ignition Residue 0.1% max.
Infrared Spectrum Authentic
Water 5% max.
Quantity 1 g
Solubility Information Solubility in water: very slightly soluble. Other solubilities: very slightly soluble in methanol, soluble in dmso, insoluble in ethanol
Formula Weight 435.43
Percent Purity ≥90% (HPLC)
Chemical Name or Material Aztreonam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.