missing translation for 'onlineSavingsMsg'
Learn More

Aztreonam, 95.7%, MP Biomedicals™

Product Code. 11403281 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403281 1 g 1g
11413281 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403281 Supplier MP Biomedicals Supplier No. 0215041501
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specifically targets penicillin-binding protein 3 (PBP-3) of Gram-negative bacteria

  • Displays high degree of resistance to β-lactamase degradation
  • With high specificity for gram-negative aerobic rods
  • White Powder
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
Synonym Azthreonam, Azactam
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Type Aztreonam
Physical Form Crystalline Powder
Color White
Quantity 1 g
Assay Percent Range 0.957
Formula Weight 435.4
Chemical Name or Material Aztreonam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.