missing translation for 'onlineSavingsMsg'
Learn More

Azure B Certified, MP Biomedicals™

Product Code. 11406051 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406051 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406051 Supplier MP Biomedicals Supplier No. 0219052010
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Azure B Certified is used as a stain.

  • Azure B Certified is used for differentiating cellular RNA (purplish) and DNA (blue-green) in plant tissues.
  • Useful as a stain for Negri Bodies.
  • Can be used in the Romanowsky-type stains for blood and blood protozoa.

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15H16ClN3S
MDL Number MFCD00011935
Synonym C.I. 52010, N, N, N-Trimethylthionin
PubChem CID 68275

Specifications

Color Green
Quantity 10 g
InChI Key DNDJEIWCTMMZBX-UHFFFAOYSA-N
Physical Form Powder
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
Molecular Weight (g/mol) 305.824
Formula Weight 305.8
Chemical Name or Material Azure B

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.