accessibility menu, dialog, popup

UserName

ClH3CNHNSNCH3CH3

CAS RN 531-55-5

IUPAC Name: 3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: 3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular Weight (g/mol): 305.82
Molecular Formula: C15H16ClN3S
InChi Key: KFZNPGQYVZZSNV-UHFFFAOYSA-M
SMILES: [Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Color 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)