missing translation for 'onlineSavingsMsg'
Learn More

Azure B (Technical Grade), TRC

Product Code. 30138609
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138609 10 g 10g
17972815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138609 Supplier Toronto Research Chemicals Supplier No. TRCA96530510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15 H16 N3 S . Cl
Molecular Weight (g/mol) 305.83
Synonym Azur I, Azure B Chloride, Azure I, C.I. 52010, Methylene Azure, Trimethylthionine Chloride, 3-Methylamino-7-dimethylaminophenazathionium Chloride, Trimethylthionine, 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium Chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ylium Chloride
IUPAC Name 3-N,7-N,7-N-trimethylphenothiazin-5-ium-3,7-diamine;chloride
SMILES [Cl-].CNc1ccc2nc3ccc(cc3[s+]c2c1)N(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H16N3S.ClH/c1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;/h4-9,16H,1-3H3;1H/q+1;/p-1
Formula Weight 305.075
Chemical Name or Material Azure B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.