missing translation for 'onlineSavingsMsg'
Learn More

Azure B (Technical Grade), TRC

Product Code. 17972815
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972815 5 g 5g
30138609 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17972815 Supplier Toronto Research Chemicals Supplier No. TRCA9653055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-55-5
Formula Weight 305.075
InChI Formula InChI=1S/C15H16N3S.ClH/c1-16-10-4-6-12-14(8-10)19-15-9-11(18(2)3)5-7-13(15)17-12;/h4-9,16H,1-3H3;1H/q+1;/p-1
IUPAC Name 3-N,7-N,7-N-trimethylphenothiazin-5-ium-3,7-diamine;chloride
Molecular Formula C15 H16 N3 S . Cl
Molecular Weight (g/mol) 305.83
SMILES [Cl-].CNc1ccc2nc3ccc(cc3[s+]c2c1)N(C)C
Synonym Azur I, Azure B Chloride, Azure I, C.I. 52010, Methylene Azure, Trimethylthionine Chloride, 3-Methylamino-7-dimethylaminophenazathionium Chloride, Trimethylthionine, 3-(Dimethylamino)-7-(methylamino)-phenothiazin-5-ium Chloride, 3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ylium Chloride
Chemical Name or Material Azure B

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.