missing translation for 'onlineSavingsMsg'
Learn More

Azure C

Product Code. 15426439
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15426439

Brand: Thermo Scientific Alfa Aesar J64515.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 531-57-7 | C13H12ClN3S | 277.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-57-7
Molecular Formula C13H12ClN3S
Molecular Weight (g/mol) 277.77
MDL Number MFCD00067677
InChI Key DDGMDTGNGDOUPX-UHFFFAOYSA-N
Synonym C.I. 52002; 3-Amino-7-(methylamino)phenothiazin-5-ium chloride
PubChem CID 68277
IUPAC Name (7-aminophenothiazin-3-ylidene)-methylazanium;chloride
SMILES C[NH+]=C1C=CC2=NC3=C(C=C(C=C3)N)SC2=C1.[Cl-]

Specifications

Beilstein 4060661
Merck Index 14,929
Formula Weight 277.78
Color Black or Green
Quantity 1 g
Physical Form Powder
Chemical Name or Material Azure C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.