missing translation for 'onlineSavingsMsg'
Learn More

Azure I

Product Code. 11335099
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335099 100 g 100g
11315099 5 g 5g
11325099 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335099 Supplier Thermo Scientific Alfa Aesar Supplier No. A17508.22
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 531-55-5 | C15H16ClN3S | 305.824 g/mol

Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.824
MDL Number MFCD00011935
InChI Key DNDJEIWCTMMZBX-UHFFFAOYSA-N
Synonym azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech
PubChem CID 68275
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Melting Point ∼205°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 3922630
Merck Index 14,6058
Solubility Information Soluble in 1mg/ml water. Sparingly soluble in alcohol.
Formula Weight 305.83
Chemical Name or Material Azure I

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.