missing translation for 'onlineSavingsMsg'
Learn More

B, Ba, Ca, Mg, Mo, Na, P, Si, Zn @ 2500 μg/g in 75cSt oil, VHG™

Product Code. 17970496
Change view
Click to view available options
Quantity:
200 g
Unit Size:
200g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970496 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970496 Supplier VHG Labs Supplier No. VHGMA92500200G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

B, Ba, Ca, Mg, Mo, Na, P, Si, Zn @ 2500 μg/g in 75cSt oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Barium/Boron/Calcium/Magnesium/Molybdenum/Phosphorus/Silicon/Sodium/Zinc
CAS 7440-39-3, 7440-42-8, 7440-70-2, 7439-95-4, 7439-98-7, 7723-14-0
Methods ICP
Analyte or Component Names Barium/Boron/Calcium/Magnesium/Molybdenum/Phosphorus/Silicon/Sodium/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix 75 cSt Oil
Quantity 200 g
Concentration or Composition (by Analyte or Components) Barium 2500 μg/g/Boron 2500 μg/g/Calcium 2500 μg/g/Magnesium 2500 μg/g/Molybdenum 2500 μg/g/Phosphorus 2500 μg/g/Silicon 2500 μg/g/Sodium 2500 μg/g/Zinc 2500 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.