missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GALT4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92226-0.02ml
This item is not returnable.
View return policy
Description
B3GALT4 Polyclonal antibody specifically detects B3GALT4 in Mouse samples. It is validated for Western Blot
Specifications
| B3GALT4 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| b3Gal-T4, beta-1,3-galactosyltransferase 4, Beta-1,3-GalTase 4, beta-3-galactosyltransferase 4, Beta3GalT4, Beta3Gal-T4, EC 2.4.1.62, Gal-T2, GALT2, GalT4, Ganglioside galactosyltransferase, UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 4, UDP-galactose:beta-N-acetyl-galactosamine-beta-1,3-galactosyltransferase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 80-270 of human B3GALT4 (NP_003773.1). APENLNQRNAIRASWGGLREARGLRVQTLFLLGEPNAQHPVWGSQGSDLASESAAQGDILQAAFQDSYRNLTLKTLSGLNWAEKHCPMARYVLKTDDDVYVNVPELVSELVLRGGRWGQWERSTEPQREAEQEGGQVLHSEEVPLLYLGRVHWRVNPSRTPGGRHRVSEEQWPHTWGPFPPYASGTGYVLS | |
| 0.02 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8705 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction