missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Antibody (62N3G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
242.00 € - 536.00 €
Specifications
| Antigen | B7-1/CD80 |
|---|---|
| Clone | 62N3G8 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot reported by customer review, Flow Cytometry 1:10-1:1000. Use reported by customer review, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18018233
|
Novus Biologicals
NBP2-25255SS |
0.025 mg |
242.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18705303
|
Novus Biologicals
NBP2-25255 |
536.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| B7-1/CD80 | |
| 1.0 mg/ml | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Cytokine Research, Immunology, Signal Transduction | |
| NP_005182 | |
| 941 | |
| Amino acids 102-223 of human CD80 were used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 62N3G8 | |
| Western Blot reported by customer review, Flow Cytometry 1:10-1:1000. Use reported by customer review, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Activation B7-1 antigen, B7, BB1, CD28LG1, CD28LGB7-1 antigen), CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
| CD80 | |
| IgG2b κ | |
| Protein G purified | |
| This product reacts with Human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title