missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-H2/ICOSLG Antibody (ICOSL/3260), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79932R
This item is not returnable.
View return policy
Description
B7-H2/ICOSLG Monoclonal specifically detects B7-H2/ICOSLG in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| B7-H2/ICOSLG | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ICOSLG | |
| Recombinant fragment of human B7-H2/ICOSLG protein (around aa 23-149) (exact sequence is proprietary) (Uniprot: O75144) | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| This monoclonal antibody recognizes a protein of 36kDa, identified as B7-H2/ICOSLG. | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| ICOSL/3260 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction